Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lush | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169997.1 | NP_001163468.1 | 3 - 4 | 580 | 65 | |
NM_079438.3 | NP_524162.1 | 3 - 4 | 593 | 65 | ||
GenBank mRNA | AF001621.1 | - | 3 - 4 | 593 | 65 | |
BT024426.1 | - | 3 - 4 | 580 | 65 | ||
KX531382.1 | - | 2 - 3 | 369 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |