Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136973.2 | NP_610817.1 | 1 - 2 | 166 | 66 | |
GenBank mRNA | BT025923.1 | - | 1 - 2 | 155 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |