Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: row | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274063.2 | NP_001260992.1 | 4 - 5 | 2770 | 76 | |
NM_137175.4 | NP_611019.2 | 4 - 5 | 2770 | 76 | ||
GenBank mRNA | BT016036.1 | - | 4 - 5 | 2749 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |