Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042796.3 | NP_001036261.3 | 23 - 24 | 8070 | 135 | |
NM_001297950.1 | NP_001284879.1 | 23 - 24 | 8412 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |