Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103427.3 | NP_001096897.1 | 13 - 14 | 3988 | 57 | |
NM_001258625.2 | NP_001245554.1 | 14 - 15 | 4021 | 57 | ||
NM_132124.5 | NP_572352.1 | 12 - 13 | 3484 | 57 | ||
NM_167091.3 | NP_727125.1 | 12 - 13 | 3445 | 57 | ||
NM_167092.3 | NP_727126.1 | 12 - 13 | 3444 | 57 | ||
NM_167093.2 | NP_727127.1 | 12 - 13 | 3384 | 57 | ||
NM_167094.2 | NP_727128.1 | 12 - 13 | 3453 | 57 | ||
GenBank mRNA | BT003254.1 | - | 12 - 13 | 3431 | 57 | |
BT124988.1 | - | 14 - 15 | 4001 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |