Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: X11Lbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272496.2 | NP_001259425.2 | 9 - 10 | 6919 | 107 | |
NM_001298156.1 | NP_001285085.1 | 11 - 12 | 6046 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |