Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299619.1 | NP_001286548.1 | 1 - 2 | 641 | 95 | |
NM_137416.3 | NP_611260.1 | 1 - 2 | 641 | 95 | ||
GenBank mRNA | AY069597.1 | - | 1 - 2 | 612 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |