Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103917.3 | NP_001097387.1 | 5 - 6 | 1368 | 68 | |
NM_001274182.2 | NP_001261111.1 | 5 - 6 | 1368 | 68 | ||
NM_001299720.1 | NP_001286649.1 | 3 - 4 | 557 | 68 | ||
NM_137678.3 | NP_611522.1 | 5 - 6 | 1368 | 68 | ||
GenBank mRNA | AY061821.1 | - | 5 - 6 | 1353 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |