Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: na | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103511.3 | NP_001096981.2 | 13 - 14 | 5719 | 89 | |
NM_167401.3 | NP_727772.3 | 13 - 14 | 5692 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |