Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201739.2 | NP_001188668.1 | 3 - 4 | 2535 | 96 | |
NM_001258798.2 | NP_001245727.1 | 5 - 6 | 2142 | 96 | ||
NM_001272735.3 | NP_001259664.1 | 3 - 4 | 2535 | 96 | ||
NM_167594.4 | NP_728122.1 | 3 - 4 | 2535 | 96 | ||
NM_206775.5 | NP_996498.2 | 5 - 6 | 2142 | 96 | ||
GenBank mRNA | X07132.1 | - | 416 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |