Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138171.2 | NP_612015.1 | 8 - 9 | 3739 | 70 | |
NM_167813.2 | NP_728501.1 | 4 - 5 | 2275 | 70 | ||
GenBank mRNA | BT010317.1 | - | 8 - 9 | 3739 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |