Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297850.1 | NP_001284779.1 | 2 - 3 | 1504 | 75 | |
NM_130559.3 | NP_569915.1 | 2 - 3 | 1504 | 75 | ||
GenBank mRNA | AY047556.1 | - | 2 - 3 | 1473 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |