Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143682.4 | NP_651939.4 | 3 - 4 | 986 | 150 | |
GenBank mRNA | BT011201.1 | - | 3 - 4 | 988 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |