Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4d14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143763.4 | NP_652020.1 | 1 - 2 | 242 | 71 | |
GenBank mRNA | BT050587.1 | - | 1 - 2 | 240 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |