Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmeg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169838.3 | NP_001163309.2 | 3 - 4 | 717 | 85 | |
NM_001169839.3 | NP_001163310.2 | 3 - 4 | 838 | 85 | ||
NM_138187.4 | NP_612031.3 | 3 - 4 | 729 | 85 | ||
NM_167830.4 | NP_728522.2 | 3 - 4 | 889 | 85 | ||
NM_167831.3 | NP_728523.2 | 4 - 5 | 926 | 85 | ||
GenBank mRNA | BT044533.1 | - | 3 - 4 | 797 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |