Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274312.1 | NP_001261241.1 | 3 - 4 | 2472 | 69 | |
NM_167852.2 | NP_728565.1 | 3 - 4 | 2475 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |