Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: promL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259658.3 | NP_001246587.1 | 6 - 7 | 1230 | 72 | |
NM_001274422.1 | NP_001261351.1 | 5 - 6 | 1147 | 72 | ||
NM_001274424.1 | NP_001261353.1 | 5 - 6 | 1349 | 72 | ||
NM_001382193.1 | NP_001368963.1 | 6 - 7 | 1230 | 72 | ||
NM_139513.3 | NP_647770.1 | 5 - 6 | 1071 | 72 | ||
NM_167986.1 | NP_728809.1 | 5 - 6 | 1349 | 72 | ||
NM_167987.4 | NP_728810.1 | 6 - 7 | 1230 | 72 | ||
GenBank mRNA | AF127935.2 | - | 6 - 7 | 1205 | 72 | |
AF197345.1 | - | 5 - 6 | 1027 | 72 | ||
BT031128.1 | - | 5 - 6 | 1029 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |