Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dikar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202125.2 | NP_001189054.1 | 7 - 8 | 3037 | 66 | |
NM_001274551.1 | NP_001261480.1 | 7 - 8 | 3037 | 66 | ||
NM_001274552.1 | NP_001261481.1 | 7 - 8 | 2611 | 66 | ||
NM_168179.3 | NP_729188.2 | 7 - 8 | 2611 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |