Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BtbVII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139517.3 | NP_647774.2 | 6 - 7 | 2731 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |