Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: armi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014556.4 | NP_001014556.1 | 4 - 5 | 1362 | 61 | |
NM_001259674.1 | NP_001246603.1 | 2 - 3 | 859 | 61 | ||
NM_139559.4 | NP_647816.2 | 4 - 5 | 1379 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |