Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259770.1 | NP_001246699.1 | 27 - 28 | 10703 | 111 | |
NM_001259771.1 | NP_001246700.1 | 26 - 27 | 10200 | 111 | ||
NM_001259772.1 | NP_001246701.1 | 27 - 28 | 10362 | 111 | ||
NM_001274708.1 | NP_001261637.1 | 25 - 26 | 10529 | 111 | ||
NM_001274709.1 | NP_001261638.1 | 27 - 28 | 10344 | 111 | ||
NM_001274710.1 | NP_001261639.1 | 26 - 27 | 10218 | 111 | ||
NM_168356.2 | NP_729519.2 | 26 - 27 | 10314 | 111 | ||
NM_168357.3 | NP_729520.1 | 25 - 26 | 10170 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |