Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168397.2 | NP_729591.1 | 4 - 5 | 797 | 54 | |
NM_168398.2 | NP_729592.1 | 4 - 5 | 720 | 54 | ||
NM_168399.2 | NP_729593.1 | 4 - 5 | 880 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |