Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104130.2 | NP_001097600.1 | 1 - 2 | 946 | 63 | |
NM_168551.2 | NP_729910.1 | 1 - 2 | 946 | 63 | ||
GenBank mRNA | AY059435.1 | - | 1 - 2 | 934 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |