Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274907.1 | NP_001261836.1 | 2 - 3 | 1510 | 61 | |
NM_080073.3 | NP_524812.1 | 2 - 3 | 1510 | 61 | ||
GenBank mRNA | AY051808.1 | - | 2 - 3 | 1478 | 61 | |
X54646.1 | - | 1507 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |