Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ergic53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080037.3 | NP_524776.1 | 1 - 2 | 209 | 59 | |
NM_176301.2 | NP_788479.1 | 1 - 2 | 209 | 59 | ||
GenBank mRNA | AF223385.1 | - | 1 - 2 | 199 | 59 | |
AY051971.1 | - | 1 - 2 | 199 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |