Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103493.4 | NP_001096963.1 | 6 - 7 | 4768 | 75 | |
NM_001103494.2 | NP_001096964.1 | 6 - 7 | 4887 | 75 | ||
NM_001298229.1 | NP_001285158.1 | 7 - 8 | 5202 | 75 | ||
NM_132584.3 | NP_572812.3 | 7 - 8 | 5076 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |