Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yippee | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038752.2 | NP_001033841.1 | 2 - 3 | 463 | 72 | |
NM_132654.4 | NP_572882.1 | 2 - 3 | 463 | 72 | ||
GenBank mRNA | AF172939.1 | - | 2 - 3 | 402 | 72 | |
AY118569.1 | - | 2 - 3 | 425 | 72 | ||
BT120317.1 | - | 2 - 3 | 281 | 72 | ||
KX531577.1 | - | 1 - 2 | 123 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |