Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272656.2 | NP_001259585.1 | 10 - 11 | 5623 | 64 | |
NM_001272657.2 | NP_001259586.1 | 10 - 11 | 5543 | 64 | ||
NM_001298376.1 | NP_001285305.1 | 10 - 11 | 5645 | 64 | ||
NM_132858.3 | NP_573086.2 | 10 - 11 | 5648 | 64 | ||
GenBank mRNA | AY118524.1 | - | 10 - 11 | 5623 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |