Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169885.2 | NP_001163356.1 | 3 - 4 | 480 | 65 | |
NM_001169887.2 | NP_001163358.1 | 3 - 4 | 485 | 65 | ||
NM_001169890.2 | NP_001163361.1 | 3 - 4 | 485 | 65 | ||
NM_001169891.2 | NP_001163362.2 | 3 - 4 | 460 | 65 | ||
NM_001169892.2 | NP_001163363.1 | 3 - 4 | 485 | 65 | ||
NM_001169893.2 | NP_001163364.1 | 3 - 4 | 485 | 65 | ||
NM_001259722.1 | NP_001246651.1 | 3 - 4 | 455 | 65 | ||
NM_001300032.1 | NP_001286961.1 | 3 - 4 | 485 | 65 | ||
NM_001300033.1 | NP_001286962.1 | 3 - 4 | 485 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |