Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr66Cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139952.3 | NP_648209.1 | 2 - 3 | 324 | 79 | |
GenBank mRNA | BT024400.1 | - | 2 - 3 | 324 | 79 | |
KX532142.1 | - | 1 - 2 | 259 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |