Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RanGAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273649.1 | NP_001260578.1 | 3 - 4 | 1419 | 64 | |
NM_001273650.1 | NP_001260579.1 | 4 - 5 | 1555 | 64 | ||
NM_057364.4 | NP_476712.1 | 4 - 5 | 1812 | 64 | ||
GenBank mRNA | AY061219.1 | - | 3 - 4 | 1404 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |