Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Spec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274357.1 | NP_001261286.1 | 8 - 9 | 7519 | 67 | |
NM_001274358.1 | NP_001261287.1 | 7 - 8 | 7489 | 67 | ||
NM_001274359.1 | NP_001261288.1 | 7 - 8 | 7423 | 67 | ||
NM_057391.4 | NP_476739.1 | 6 - 7 | 7393 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |