Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299844.1 | NP_001286773.1 | 6 - 7 | 1480 | 82 | |
NM_001299845.1 | NP_001286774.1 | 2 - 3 | 246 | 82 | ||
NM_001299846.1 | NP_001286775.1 | 2 - 3 | 730 | 82 | ||
NM_057516.4 | NP_476864.2 | 6 - 7 | 1486 | 82 | ||
NM_176254.2 | NP_788434.1 | 6 - 7 | 1501 | 82 | ||
GenBank mRNA | AY118955.1 | - | 6 - 7 | 1470 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |