Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bowl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258932.2 | NP_001245861.1 | 3 - 4 | 1283 | 70 | |
NM_001258933.2 | NP_001245862.1 | 3 - 4 | 1170 | 70 | ||
NM_001258934.1 | NP_001245863.1 | 3 - 4 | 1211 | 70 | ||
NM_057535.4 | NP_476883.1 | 3 - 4 | 1200 | 70 | ||
NM_164556.2 | NP_722939.1 | 2 - 3 | 1261 | 70 | ||
NM_164557.3 | NP_722940.1 | 3 - 4 | 1289 | 70 | ||
NM_164558.2 | NP_722941.1 | 3 - 4 | 1406 | 70 | ||
GenBank mRNA | BT010294.1 | - | 3 - 4 | 1407 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |