Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057539.5 | NP_476887.2 | 2 - 3 | 735 | 74 | |
NM_164834.2 | NP_723414.2 | 2 - 3 | 1304 | 74 | ||
GenBank mRNA | BT003533.1 | - | 2 - 3 | 731 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |