Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080523.4 | NP_536784.1 | 1 - 2 | 171 | 84 | |
GenBank mRNA | AF233526.1 | - | 1 - 2 | 123 | 84 | |
AY051959.1 | - | 1 - 2 | 149 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |