Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058037.5 | NP_477385.1 | 1 - 2 | 201 | 78 | |
GenBank mRNA | AF132551.1 | - | 1 - 2 | 230 | 78 | |
U89162.1 | - | 1 - 2 | 167 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |