Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058054.4 | NP_477402.1 | 1 - 2 | 520 | 78 | |
GenBank mRNA | AF111427.1 | - | 1 - 2 | 179 | 78 | |
BT012453.1 | - | 1 - 2 | 518 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |