Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259620.2 | NP_001246549.1 | 3 - 4 | 980 | 90 | |
NM_001274346.1 | NP_001261275.1 | 3 - 4 | 980 | 90 | ||
NM_001274347.1 | NP_001261276.1 | 3 - 4 | 980 | 90 | ||
NM_001274348.1 | NP_001261277.1 | 3 - 4 | 980 | 90 | ||
NM_058102.3 | NP_477450.1 | 3 - 4 | 980 | 90 | ||
NM_167911.3 | NP_728660.2 | 3 - 4 | 980 | 90 | ||
GenBank mRNA | BT044170.1 | - | 3 - 4 | 966 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |