Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176272.2 | NP_788450.1 | 6 - 7 | 881 | 104 | |
NM_176273.2 | NP_788451.1 | 2 - 3 | 399 | 104 | ||
GenBank mRNA | AY119150.1 | - | 2 - 3 | 386 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |