Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tacc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202238.2 | NP_001189167.1 | 6 - 7 | 758 | 89 | |
NM_001202239.2 | NP_001189168.1 | 8 - 9 | 3471 | 89 | ||
NM_001202240.2 | NP_001189169.1 | 8 - 9 | 3865 | 89 | ||
NM_001259986.1 | NP_001246915.1 | 8 - 9 | 3392 | 89 | ||
NM_001275332.1 | NP_001262261.1 | 8 - 9 | 3865 | 89 | ||
NM_001275333.1 | NP_001262262.1 | 9 - 10 | 3706 | 89 | ||
NM_058085.5 | NP_477433.2 | 8 - 9 | 3823 | 89 | ||
NM_058086.4 | NP_477434.1 | 8 - 9 | 3510 | 89 | ||
NM_169016.3 | NP_730867.2 | 8 - 9 | 3513 | 89 | ||
NM_176401.2 | NP_788578.1 | 8 - 9 | 3778 | 89 | ||
NM_176402.3 | NP_788579.1 | 6 - 7 | 716 | 89 | ||
NM_176403.2 | NP_788580.1 | 6 - 7 | 709 | 89 | ||
NM_176404.2 | NP_788581.1 | 6 - 7 | 751 | 89 | ||
GenBank mRNA | AY058499.1 | - | 8 - 9 | 3823 | 89 | |
BT099701.1 | - | 8 - 9 | 3479 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |