Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sodh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079587.3 | NP_524311.1 | 1 - 2 | 126 | 60 | |
GenBank mRNA | AY058731.1 | - | 1 - 2 | 114 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |