Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260137.1 | NP_001247066.1 | 6 - 7 | 1364 | 63 | |
NM_079600.3 | NP_524324.2 | 7 - 8 | 1694 | 63 | ||
NM_169444.1 | NP_731661.1 | 7 - 8 | 1382 | 63 | ||
NM_176466.1 | NP_788643.1 | 7 - 8 | 1401 | 63 | ||
NM_176467.1 | NP_788644.1 | 7 - 8 | 1379 | 63 | ||
NM_176468.1 | NP_788645.1 | 6 - 7 | 1778 | 63 | ||
NM_176469.2 | NP_788646.1 | 7 - 8 | 1499 | 63 | ||
NM_176470.1 | NP_788647.1 | 7 - 8 | 1404 | 63 | ||
NM_176471.1 | NP_788648.1 | 7 - 8 | 1577 | 63 | ||
NM_176472.1 | NP_788649.1 | 7 - 8 | 1599 | 63 | ||
NM_176473.1 | NP_788650.1 | 8 - 9 | 1473 | 63 | ||
NM_176474.1 | NP_788651.1 | 8 - 9 | 1451 | 63 | ||
NM_176475.1 | NP_788652.1 | 7 - 8 | 1850 | 63 | ||
NM_176476.1 | NP_788653.1 | 8 - 9 | 1454 | 63 | ||
NM_176477.1 | NP_788654.1 | 8 - 9 | 1476 | 63 | ||
NM_176478.1 | NP_788655.1 | 8 - 9 | 1649 | 63 | ||
GenBank mRNA | AY069094.1 | - | 7 - 8 | 1364 | 63 | |
BT120190.1 | - | 6 - 7 | 1364 | 63 | ||
BT125878.1 | - | 8 - 9 | 1450 | 63 | ||
BT133296.1 | - | 8 - 9 | 1628 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |