Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079644.3 | NP_524368.5 | 2 - 3 | 1107 | 60 | |
GenBank mRNA | Z86118.1 | - | 1111 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |