Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104392.3 | NP_001097862.1 | 9 - 10 | 2385 | 58 | |
NM_001104393.3 | NP_001097863.1 | 9 - 10 | 954 | 58 | ||
NM_001275882.1 | NP_001262811.1 | 13 - 14 | 2496 | 58 | ||
NM_001382187.1 | NP_001369027.1 | 6 - 7 | 1695 | 58 | ||
NM_079717.5 | NP_524441.2 | 13 - 14 | 2496 | 58 | ||
NM_169970.3 | NP_732661.1 | 8 - 9 | 2480 | 58 | ||
GenBank mRNA | BT044550.1 | - | 9 - 10 | 2345 | 58 | |
BT046172.1 | - | 13 - 14 | 2480 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |