Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141477.2 | NP_649734.2 | 2 - 3 | 676 | 116 | |
NM_169192.3 | NP_731174.1 | 1 - 2 | 243 | 116 | ||
NM_169193.3 | NP_731175.2 | 1 - 2 | 243 | 116 | ||
GenBank mRNA | AF233093.1 | - | 1 - 2 | 234 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |