Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbp95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141503.4 | NP_649760.1 | 1 - 2 | 514 | 112 | |
GenBank mRNA | AY061240.1 | - | 1 - 2 | 481 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |