Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141983.3 | NP_650240.1 | 2 - 3 | 1226 | 59 | |
NM_169482.1 | NP_731749.3 | 3 - 4 | 1503 | 59 | ||
GenBank mRNA | AF218811.1 | - | 2 - 3 | 1195 | 59 | |
BT100045.1 | - | 2 - 3 | 1158 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |