Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MetRS-m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275635.2 | NP_001262564.1 | 2 - 3 | 465 | 64 | |
NM_001316578.1 | NP_001303507.1 | 2 - 3 | 465 | 64 | ||
NM_142091.5 | NP_650348.1 | 2 - 3 | 465 | 64 | ||
GenBank mRNA | AY069128.1 | - | 2 - 3 | 436 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |