Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HtrA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275636.1 | NP_001262565.1 | 2 - 3 | 1414 | 88 | |
NM_142109.2 | NP_650366.1 | 2 - 3 | 1162 | 88 | ||
GenBank mRNA | AB112473.1 | - | 2 - 3 | 1108 | 88 | |
AY075206.1 | - | 2 - 3 | 1162 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |